Please login to your online account to display your discounted pricing

Mouse anti-CD44, Clone: 156-3C11, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23432705ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23432705

  • / Each
 

Description & Specifications

Specifications

Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 960
Regulatory Status RUO
Gene Accession No. P16070
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Antigen CD44
Host Species Mouse
Concentration 0.2mg/mL
Purification Method Purified
Primary or Secondary Primary
Immunogen Stimulated human leukocytes
Formulation PBS with 0.05% BSA.
Cross Reactivity Baboon
Cross Reactivity Human
Cross Reactivity Monkey
Cross Reactivity Primate
Quantity 0.5mL
Format Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Reconstitution Protein A purified
Conjugate Unlabeled
Clone 156-3C11
Isotype IgG2a, κ