Please login to your online account to display your discounted pricing

Mouse anti-CD44, Clone: 156-3C11, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23432701ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23432701

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Baboon
Cross Reactivity Human
Cross Reactivity Monkey
Cross Reactivity Primate
Antigen CD44
Format Purified
Formulation PBS with 0.05% BSA.
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen Stimulated human leukocytes
Isotype IgG2a, κ
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 960
Gene Accession No. P16070
Host Species Mouse
Purification Method Purified
Quantity 0.1mL
Regulatory Status RUO
Reconstitution Protein A purified
Clone 156-3C11
Concentration 0.2mg/mL
Conjugate Unlabeled