Please login to your online account to display your discounted pricing

Mouse anti-CD43/Sialophorin, Clone: SPM503, Novus Biologicals™

$139.00 - $429.00

Specifications

Antigen CD43/Sialophorin
Clone SPM503
Concentration 0.2mg/mL
Immunogen Myeloblastic KG1 cells
Gene Accession No. P16150
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23282201

 
novus biologicals canada
NBP23282201ML
0.1mL Each for $139.00

NBP23282210

 
novus biologicals canada
NBP23282210ML
1.0mL Each for $429.00
Description & Specifications

Specifications

Antigen CD43/Sialophorin
Clone SPM503
Concentration 0.2mg/mL
Immunogen Myeloblastic KG1 cells
Gene Accession No. P16150
Gene ID (Entrez) 6693
Formulation PBS with 0.05% BSA.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting