missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mouse anti-CD43/Sialophorin, Clone: SPM503, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen CD43/Sialophorin
Clone SPM503
Concentration 1mg/mL
Immunogen Myeloblastic KG1 cells
Gene Accession No. P16150
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP23477501 Novus Biologicals Canada
NBP23477501MG
0.1mg Each for $384.00
NBP23477502 Novus Biologicals Canada
NBP23477502MG
0.2mg Each for $527.00
Specifications Description & Specifications

Specifications

CD43/Sialophorin
1mg/mL
P16150
10mM PBS. Does not contain BSA. Azide Free.
Primary
Murine
Unlabeled
Purified
SPM503
Myeloblastic KG1 cells
6693
Monoclonal
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG1, κ
Purified
ELISA,Flow Cytometry,Immunocytochemistry,Immunofluorescence,Immunohistochemistry (Frozen),Immunohistochemistry (Paraffin),Immunoprecipitation,Western Blot