Please login to your online account to display your discounted pricing

Mouse anti-CD43/Sialophorin, Clone: SPM503, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23477502MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23477502

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Regulatory Status RUO
Host Species Mouse
Monoclonal or Polyclonal Monoclonal
Antigen CD43/Sialophorin
Conjugate Unlabeled
Isotype IgG1, κ
Gene Accession No. P16150
Quantity 0.2mg
Primary or Secondary Primary
Reconstitution Protein A purified
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Purification Method Purified
Clone SPM503
Test Specificity It recognizes a cell surface glycoprotein of 95/115/135kDa (depending upon the extent of glycosylation), identified as CD43. 70-90% of T-cell lymphomas and from 22-37% of B-cell lymphomas express CD43.
Cross Reactivity Human
Format Purified
Gene ID (Entrez) 6693
Concentration 1mg/mL
Immunogen Myeloblastic KG1 cells