Please login to your online account to display your discounted pricing

Mouse anti-CD43/Sialophorin, Clone: DF-T1, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP21519002MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP21519002

  • / Each
 

Description & Specifications

Specifications

Gene Accession No. P16150
Concentration 0.2mg/mL
Antigen CD43/Sialophorin
Immunogen Myeloblastic KG1 cells were used as the immunogen for this antibody.
Formulation PBS with 0.05% BSA. Protein G purified.
Gene ID (Entrez) 6693
Format Purified
Test Specificity It recognizes a cell surface glycoprotein of 95/115/135kDa (depending upon the extent of glycosylation), identified as CD43. 70-90% of T-cell lymphomas and from 22-37% of B-cell lymphomas express CD43. No reactivity has been observed with reactive B-cells. So a B-lineage population that co-expresses CD43 is highly likely to be a malignant lymphoma, especially a low-grade lymphoma, rather than a reactive B-cell population. When CD43 antibody is used in combination with anti-CD20, effective immunophenotyping of the lymphomas in formalin-fixed tissues can be obtained. Co-staining of a lymphoid infiltrate with anti-CD20 and anti-CD43 argues against a reactive process and favors a diagnosis of lymphoma.
Monoclonal or Polyclonal Monoclonal
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Conjugate Unlabeled
Host Species Mouse
Isotype IgG1, κ
Primary or Secondary Primary
Regulatory Status RUO
Quantity 0.2mg
Purification Method Purified
Clone DF-T1
Cross Reactivity Human
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.