Please login to your online account to display your discounted pricing

Mouse anti-CD40/TNFRSF5, Clone: 5C3, PE, Novus Biologicals™

$131.00 - $296.00

Specifications

Antigen CD40/TNFRSF5
Clone 5C3
Immunogen The immunogen for this antibody was CD40.
Gene ID (Entrez) 958
Formulation Phosphate buffer pH 7.2
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP143869

 
novus biologicals canada
NBP143869100TESTS
100 Tests Each for $296.00

NBP14386925

 
novus biologicals canada
NBP14386925TESTS
25 Tests Each for $131.00
Description & Specifications

Specifications

Antigen CD40/TNFRSF5
Clone 5C3
Immunogen The immunogen for this antibody was CD40.
Gene ID (Entrez) 958
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1, κ
Conjugate PE
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry