Please login to your online account to display your discounted pricing

Mouse anti-CD40/TNFRSF5, Clone: 5C3, FITC, Novus Biologicals™

$131.00 - $252.00

Specifications

Antigen CD40/TNFRSF5
Clone 5C3
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CD40.
Gene ID (Entrez) 958
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP144013

 
novus biologicals canada
NBP144013100TESTS
100 Tests Each for $252.00

NBP14401325

 
novus biologicals canada
NBP14401325TESTS
25 Tests Each for $131.00
Description & Specifications

Specifications

Antigen CD40/TNFRSF5
Clone 5C3
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CD40.
Gene ID (Entrez) 958
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Murine
Isotype IgG1, κ
Conjugate FITC
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry