Please login to your online account to display your discounted pricing

Mouse anti-CD40 Ligand/TNFSF5, Clone: 24-31, PE, Novus Biologicals™

$171.00 - $354.00

Specifications

Antigen CD40 Ligand/TNFSF5
Clone 24-31
Immunogen The immunogen was the full length version of the protein: CD154 (24-31)
Gene Accession No. P29965
Gene ID (Entrez) 959
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP200182

 
novus biologicals canada
NBP200182100TESTS
100 Tests Each for $354.00

NBP20018225

 
novus biologicals canada
NBP20018225TESTS
25 Tests Each for $171.00
Description & Specifications

Specifications

Antigen CD40 Ligand/TNFSF5
Clone 24-31
Immunogen The immunogen was the full length version of the protein: CD154 (24-31)
Gene Accession No. P29965
Gene ID (Entrez) 959
Formulation Immunogen affinity purified
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C in the dark.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1, κ
Conjugate PE
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry