Please login to your online account to display your discounted pricing

Mouse anti-CD40 Ligand/TNFSF5, Clone: 24-31, PE, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP20018225TESTS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP20018225

  • / Each
 

Description & Specifications

Specifications

Antigen CD40 Ligand/TNFSF5
Applications Flow Cytometry
Clone 24-31
Conjugate PE
Cross Reactivity Human
Format Affinity Purified
Formulation Immunogen affinity purified
Gene Accession No. P29965
Host Species Mouse
Immunogen The immunogen was the full length version of the protein: CD154 (24-31)
Isotype IgG1, κ
Purification Method Affinity Purified
Quantity 25 Tests
Regulatory Status RUO
Storage Requirements Store at 4°C in the dark.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 959
Test Specificity CD154 (24-31). Expected Cross-Reactivity based on immunogen sequence: Cynomolgus, Rhesus.