Please login to your online account to display your discounted pricing

Mouse anti-CD40 Ligand/TNFSF5, Clone: 24-31, PE, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP200182100TESTS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP200182

  • / Each
 

Description & Specifications

Specifications

Host Species Mouse
Monoclonal or Polyclonal Monoclonal
Antigen CD40 Ligand/TNFSF5
Regulatory Status RUO
Immunogen The immunogen was the full length version of the protein: CD154 (24-31)
Gene Accession No. P29965
Applications Flow Cytometry
Quantity 100 Tests
Cross Reactivity Human
Conjugate PE
Formulation Immunogen affinity purified
Test Specificity CD154 (24-31). Expected Cross-Reactivity based on immunogen sequence: Cynomolgus, Rhesus.
Format Affinity Purified
Isotype IgG1, κ
Purification Method Affinity Purified
Storage Requirements Store at 4°C in the dark.
Clone 24-31
Primary or Secondary Primary
Gene ID (Entrez) 959