Please login to your online account to display your discounted pricing

Mouse anti-CD40 Ligand/TNFSF5, Clone: 24-31, APC, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP200074100TESTS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP200074

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Host Species Mouse
Primary or Secondary Primary
Antigen CD40 Ligand/TNFSF5
Regulatory Status RUO
Gene ID (Entrez) 959
Clone 24-31
Quantity 100 Tests
Conjugate APC
Purification Method Affinity Purified
Formulation Immunogen affinity purified
Monoclonal or Polyclonal Monoclonal
Storage Requirements Store at 4°C. Do not freeze.
Gene Accession No. P29965
Immunogen The immunogen was the full length version of the protein: CD154 (24-31)
Cross Reactivity Human
Format Affinity Purified
Isotype IgG1, κ
Test Specificity CD154 (24-31). Expected Cross-Reactivity based on immunogen sequence: Cynomolgus, Rhesus.