Please login to your online account to display your discounted pricing

Mouse anti-CD40 Ligand/TNFSF5, Clone: 24-31, APC, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP20007425TESTS

 View all options for this product


For Research Use Only

Catalog No. NBP20007425


Description & Specifications


Antigen CD40 Ligand/TNFSF5
Applications Flow Cytometry
Clone 24-31
Conjugate APC
Cross Reactivity Human
Format Affinity Purified
Formulation Immunogen affinity purified
Gene Accession No. P29965
Host Species Mouse
Immunogen The immunogen was the full length version of the protein: CD154 (24-31)
Isotype IgG1, κ
Purification Method Affinity Purified
Quantity 25 Tests
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 959
Test Specificity CD154 (24-31). Expected Cross-Reactivity based on immunogen sequence: Cynomolgus, Rhesus.