missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mouse anti-CD39/ENTPD1, Clone: AC2.5, Novus Biologicals™

$309.05 - $332.66

Specifications

Antigen CD39/ENTPD1
Clone AC2.5
Concentration 0.5mg/mL
Immunogen CD39
Gene ID (Entrez) 953
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
DDX0321P05 Novus Biologicals Canada
DDX0321P100005MG
0.05mg Each for $309.05
DDX0321B05 Novus Biologicals Canada
DDX0321B100005MG
0.05mg Each for $332.66
Specifications Description & Specifications

Specifications

CD39/ENTPD1
0.5mg/mL
953
Monoclonal
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG1
Purified
Flow Cytometry,Immunohistochemistry (Frozen),Immunoprecipitation,Western Blot
AC2.5
CD39
Tris-NaCl pH 8
Primary
Murine
Unlabeled
Ion Exchange Chromatography