Please login to your online account to display your discounted pricing

Mouse anti-CD39/ENTPD1, Clone: AC2.5, Novus Biologicals™


Manufacturer: novus biologicals canada  DDX0321P100005MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. DDX0321P05

  • / Each
 

Description & Specifications

Specifications

Antigen CD39/ENTPD1
Host Species Mouse
Applications Flow Cytometry
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Western Blotting
Monoclonal or Polyclonal Monoclonal
Format Purified
Formulation Tris-NaCl pH 8
Isotype IgG1
Purification Method Ion Exchange Chromatography
Quantity 0.05mg
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Gene ID (Entrez) 953
Clone AC2.5
Concentration 0.5mg/mL
Conjugate Unlabeled
Immunogen CD39
Cross Reactivity Human