Please login to your online account to display your discounted pricing

Mouse anti-CD39/ENTPD1, Clone: AC2.5, Novus Biologicals™


Manufacturer: novus biologicals canada  DDX0321B100005MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. DDX0321B05

  • / Each
 

Description & Specifications

Specifications

Applications Western Blotting
Immunogen CD39
Primary or Secondary Primary
Cross Reactivity Human
Antigen CD39/ENTPD1
Formulation Tris-NaCl pH 8
Host Species Mouse
Isotype IgG1
Purification Method Ion Exchange Chromatography
Quantity 0.05mg
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 953
Concentration 0.5mg/mL
Conjugate Unlabeled
Format Purified
Clone AC2.5