Please login to your online account to display your discounted pricing

Mouse anti-CD38, Clone: HIT2, PE-Cy7, Novus Biologicals™

$197.00 - $362.00

Specifications

Antigen CD38
Clone HIT2
Immunogen Tissue / cell preparation (Human thymocytes in foetus).
Gene ID (Entrez) 952
Monoclonal or Polyclonal Monoclonal
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP200072

 
novus biologicals canada
NBP200072100TESTS
100 Tests Each for $362.00

NBP20007225

 
novus biologicals canada
NBP20007225TESTS
25 Tests Each for $197.00
Description & Specifications

Specifications

Antigen CD38
Clone HIT2
Immunogen Tissue / cell preparation (Human thymocytes in foetus).
Gene ID (Entrez) 952
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Murine
Isotype IgG1
Conjugate PE-Cy7
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry