Please login to your online account to display your discounted pricing

Mouse anti-CD35, Clone: SPM554, Novus Biologicals™

$139.00 - $429.00

Specifications

Antigen CD35
Clone SPM554
Concentration 0.2mg/mL
Immunogen Intact human monocytes
Gene Accession No. P17927
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23282101

 
novus biologicals canada
NBP23282101ML
0.1mL Each for $139.00

NBP23282110

 
novus biologicals canada
NBP23282110ML
1.0mL Each for $429.00
Description & Specifications

Specifications

Antigen CD35
Clone SPM554
Concentration 0.2mg/mL
Immunogen Intact human monocytes
Gene Accession No. P17927
Gene ID (Entrez) 1378
Formulation PBS with 0.05% BSA.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Baboon
Cross Reactivity Cynomolgus
Cross Reactivity Human
Cross Reactivity Monkey
Cross Reactivity Primate
Cross Reactivity Rhesus Ma
Isotype IgG1, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting