Please login to your online account to display your discounted pricing

Mouse anti-CD35, Clone: SPM554, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP23282110ML

 View all options for this product


For Research Use Only

Catalog No. NBP23282110


Description & Specifications


Antigen CD35
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Clone SPM554
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Baboon
Cross Reactivity Cynomolgus
Cross Reactivity Human
Cross Reactivity Monkey
Cross Reactivity Primate
Cross Reactivity Rhesus Ma
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. P17927
Host Species Mouse
Immunogen Intact human monocytes
Isotype IgG1, κ
Purification Method Purified
Quantity 1.0mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 1378
Reconstitution Protein A purified