Please login to your online account to display your discounted pricing

Mouse anti-CD35, Clone: SPM554, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23282101ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23282101

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Baboon
Cross Reactivity Cynomolgus
Cross Reactivity Human
Cross Reactivity Monkey
Cross Reactivity Primate
Cross Reactivity Rhesus Ma
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen Intact human monocytes
Monoclonal or Polyclonal Monoclonal
Clone SPM554
Quantity 0.1mL
Format Purified
Primary or Secondary Primary
Conjugate Unlabeled
Regulatory Status RUO
Reconstitution Protein A purified
Formulation PBS with 0.05% BSA.
Isotype IgG1, κ
Host Species Mouse
Antigen CD35
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Gene ID (Entrez) 1378
Concentration 0.2mg/mL
Gene Accession No. P17927
Purification Method Purified