Please login to your online account to display your discounted pricing

Mouse anti-CD35, Clone: SPM554, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23479602MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23479602

  • / Each
 

Description & Specifications

Specifications

Antigen CD35
Isotype IgG1, κ
Monoclonal or Polyclonal Monoclonal
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Primary or Secondary Primary
Immunogen Intact human monocytes
Cross Reactivity Baboon
Cross Reactivity Cynomolgus
Cross Reactivity Human
Cross Reactivity Monkey
Cross Reactivity Primate
Cross Reactivity Rhesus Ma
Conjugate Unlabeled
Quantity 0.2mg
Regulatory Status RUO
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Host Species Mouse
Test Specificity Recognizes a protein of 210-220kDa, which is identified as the complement receptor 1 (CR1)/CD35. This MAb is specific for a site in CR1 that is distal from the C3b-binding site, so that it is unable to block CR1 activity. This MAb is highly specific to CR1 and shows no cross-reaction with CR2.
Gene ID (Entrez) 1378
Concentration 1mg/mL
Clone SPM554
Purification Method Purified
Reconstitution Protein A purified
Gene Accession No. P17927
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.