Please login to your online account to display your discounted pricing

Mouse anti-CD34, Clone: SPM123, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP23293310ML

 View all options for this product


For Research Use Only

Catalog No. NBP23293310


Description & Specifications


Antigen CD34
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Clone SPM123
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Cynomolgus
Cross Reactivity Human
Cross Reactivity Monkey
Cross Reactivity Primate
Cross Reactivity Rhesus Ma
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. P28906
Host Species Mouse
Immunogen Detergent solubilized vesicular suspension prepared from human term placenta
Isotype IgG1, κ
Purification Method Purified
Quantity 1.0mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 947
Test Specificity Bovine (Negative), Canine (Negative), Rat (Negative), Sheep (Negative)
Reconstitution Protein A purified