Please login to your online account to display your discounted pricing

Mouse anti-CD34, Clone: SPM123, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23293301ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23293301

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Cross Reactivity Cynomolgus
Cross Reactivity Human
Cross Reactivity Monkey
Cross Reactivity Primate
Cross Reactivity Rhesus Ma
Antigen CD34
Format Purified
Immunogen Detergent solubilized vesicular suspension prepared from human term placenta
Purification Method Purified
Quantity 0.1mL
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 947
Test Specificity Bovine (Negative), Canine (Negative), Rat (Negative), Sheep (Negative)
Conjugate Unlabeled
Formulation PBS with 0.05% BSA.
Gene Accession No. P28906
Host Species Mouse
Isotype IgG1, κ
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Reconstitution Protein A purified
Clone SPM123
Concentration 0.2mg/mL