Please login to your online account to display your discounted pricing

Mouse anti-CD34, Clone: SPM123, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23473302MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23473302

  • / Each
 

Description & Specifications

Specifications

Immunogen Detergent solubilized vesicular suspension prepared from human term placenta
Gene ID (Entrez) 947
Cross Reactivity Human
Cross Reactivity Primate
Antigen CD34
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Gene Accession No. P28906
Host Species Mouse
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Isotype IgG1, κ
Purification Method Purified
Quantity 0.2mg
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Reconstitution Protein A purified
Clone SPM123
Conjugate Unlabeled
Test Specificity This MAb recognizes a single chain, transmembrane, heavily glycosylated protein of 90-120kDa, which is identified as CD34.
Concentration 1mg/mL