Please login to your online account to display your discounted pricing

Mouse anti-CD34, Clone: QBEnd/10, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen CD34
Clone QBEnd/10
Concentration 1mg/mL
Immunogen Detergent solubilized vesicular suspension prepared from human term placenta
Gene Accession No. P28906
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23471301

 
novus biologicals canada
NBP23471301MG
0.1mg Each for $384.00

NBP23471302

 
novus biologicals canada
NBP23471302MG
0.2mg Each for $527.00
Description & Specifications

Specifications

Antigen CD34
Clone QBEnd/10
Concentration 1mg/mL
Immunogen Detergent solubilized vesicular suspension prepared from human term placenta
Gene Accession No. P28906
Gene ID (Entrez) 947
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Cynomolgus
Cross Reactivity Human
Cross Reactivity Monkey
Cross Reactivity Primate
Cross Reactivity Rhesus Ma
Isotype IgG1, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting