Please login to your online account to display your discounted pricing

Mouse anti-CD34, Clone: QBEnd/10, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23471302MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23471302

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Primary or Secondary Primary
Quantity 0.2mg
Host Species Mouse
Immunogen Detergent solubilized vesicular suspension prepared from human term placenta
Conjugate Unlabeled
Isotype IgG1, κ
Cross Reactivity Cynomolgus
Cross Reactivity Human
Cross Reactivity Monkey
Cross Reactivity Primate
Cross Reactivity Rhesus Ma
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Concentration 1mg/mL
Regulatory Status RUO
Purification Method Purified
Gene Accession No. P28906
Format Purified
Monoclonal or Polyclonal Monoclonal
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Clone QBEnd/10
Reconstitution Protein A purified
Test Specificity Bovine (Negative), Canine (Negative), Rat (Negative), Sheep (Negative)
Gene ID (Entrez) 947
Antigen CD34