Please login to your online account to display your discounted pricing

Mouse anti-CD34, Clone: QBEnd/10, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23471301MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23471301

  • / Each
 

Description & Specifications

Specifications

Immunogen Detergent solubilized vesicular suspension prepared from human term placenta
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Clone QBEnd/10
Purification Method Purified
Format Purified
Regulatory Status RUO
Gene Accession No. P28906
Cross Reactivity Cynomolgus
Cross Reactivity Human
Cross Reactivity Monkey
Cross Reactivity Primate
Cross Reactivity Rhesus Ma
Quantity 0.1mg
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Concentration 1mg/mL
Test Specificity Bovine (Negative), Canine (Negative), Rat (Negative), Sheep (Negative)
Conjugate Unlabeled
Host Species Mouse
Antigen CD34
Gene ID (Entrez) 947
Reconstitution Protein A purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Isotype IgG1, κ