Please login to your online account to display your discounted pricing

Mouse anti-CD31/PECAM-1, Clone: SPM532, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23478101MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23478101

  • / Each
 

Description & Specifications

Specifications

Immunogen Human recombinant CD31 protein
Cross Reactivity Human
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Format Purified
Primary or Secondary Primary
Conjugate Unlabeled
Host Species Mouse
Clone SPM532
Concentration 1mg/mL
Antigen CD31/PECAM-1
Purification Method Purified
Regulatory Status RUO
Quantity 0.1mg
Isotype IgG1, κ
Monoclonal or Polyclonal Monoclonal
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Gene Accession No. P16284
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Reconstitution Protein A purified
Gene ID (Entrez) 5175