Please login to your online account to display your discounted pricing

Mouse anti-CD31/PECAM-1, Clone: SPM122, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23293001ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23293001

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Cynomolgus
Cross Reactivity Human
Cross Reactivity Monkey
Cross Reactivity Rabbit
Monoclonal or Polyclonal Monoclonal
Immunogen Membrane preparation of a spleen from a patient with hairy cell leukemia (JC/70A)
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Host Species Mouse
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Reconstitution Protein A purified
Quantity 0.1mL
Isotype IgG1, κ
Clone SPM122
Formulation PBS with 0.05% BSA.
Regulatory Status RUO
Conjugate Unlabeled
Gene ID (Entrez) 5175
Antigen CD31/PECAM-1
Concentration 0.2mg/mL
Primary or Secondary Primary
Purification Method Purified
Format Purified
Test Specificity Porcine (Negative), Rat (Negative)
Gene Accession No. P16284