Please login to your online account to display your discounted pricing

Mouse anti-CD31/PECAM-1, Clone: SPM122, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23293010ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23293010

  • / Each
 

Description & Specifications

Specifications

Quantity 1.0mL
Cross Reactivity Cynomolgus
Cross Reactivity Human
Cross Reactivity Monkey
Cross Reactivity Rabbit
Host Species Mouse
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Formulation PBS with 0.05% BSA.
Isotype IgG1, κ
Regulatory Status RUO
Gene Accession No. P16284
Immunogen Membrane preparation of a spleen from a patient with hairy cell leukemia (JC/70A)
Monoclonal or Polyclonal Monoclonal
Reconstitution Protein A purified
Concentration 0.2mg/mL
Purification Method Purified
Conjugate Unlabeled
Format Purified
Antigen CD31/PECAM-1
Gene ID (Entrez) 5175
Primary or Secondary Primary
Test Specificity Porcine (Negative), Rat (Negative)
Clone SPM122
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.