Please login to your online account to display your discounted pricing

Mouse anti-CD31/PECAM-1, Clone: SPM122, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23473202MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23473202

  • / Each
 

Description & Specifications

Specifications

Format Purified
Antigen CD31/PECAM-1
Host Species Mouse
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Gene Accession No. P16284
Quantity 0.2mg
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Gene ID (Entrez) 5175
Reconstitution Protein A purified
Cross Reactivity Cynomolgus
Cross Reactivity Human
Cross Reactivity Monkey
Cross Reactivity Rabbit
Concentration 1mg/mL
Conjugate Unlabeled
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity MAb reacts with normal, benign, and malignant endothelial cells which make up blood vessel lining. Porcine (Negative), Rat (Negative)
Clone SPM122
Isotype IgG1, κ
Immunogen Membrane preparation of a spleen from a patient with hairy cell leukemia (JC/70A)
Monoclonal or Polyclonal Monoclonal
Purification Method Purified