Please login to your online account to display your discounted pricing

Mouse anti-CD31/PECAM-1, Clone: SPM122, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23473201MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23473201

  • / Each
 

Description & Specifications

Specifications

Monoclonal or Polyclonal Monoclonal
Format Purified
Gene ID (Entrez) 5175
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Conjugate Unlabeled
Primary or Secondary Primary
Isotype IgG1, κ
Cross Reactivity Cynomolgus
Cross Reactivity Human
Cross Reactivity Monkey
Cross Reactivity Rabbit
Gene Accession No. P16284
Regulatory Status RUO
Antigen CD31/PECAM-1
Reconstitution Protein A purified
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Immunogen Membrane preparation of a spleen from a patient with hairy cell leukemia (JC/70A)
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Purification Method Purified
Host Species Mouse
Quantity 0.1mg
Clone SPM122
Concentration 1mg/mL
Test Specificity MAb reacts with normal, benign, and malignant endothelial cells which make up blood vessel lining. Porcine (Negative), Rat (Negative)