Please login to your online account to display your discounted pricing

Mouse anti-CD31/PECAM-1, Clone: JC/70A, Azide Free, Novus Biologicals™

$375.00 - $569.00

Specifications

Antigen CD31/PECAM-1
Clone JC/70A
Concentration 1mg/mL
Immunogen Membrane preparation of a spleen from a patient with hairy cell leukemia (JC/70A)
Gene Accession No. P16284
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23465102

 
novus biologicals canada
NBP23465102MG
0.2mg Each for $569.00

NBP23465101

 
novus biologicals canada
NBP23465101MG
0.1mg Each for $375.00
Description & Specifications

Specifications

Antigen CD31/PECAM-1
Clone JC/70A
Concentration 1mg/mL
Immunogen Membrane preparation of a spleen from a patient with hairy cell leukemia (JC/70A)
Gene Accession No. P16284
Gene ID (Entrez) 5175
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at -20 to -80°C. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Cynomolgus
Cross Reactivity Human
Cross Reactivity Monkey
Cross Reactivity Rabbit
Isotype IgG1, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting