missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mouse anti-CD31/PECAM-1, Clone: C31.3, Novus Biologicals™

$329.00 - $472.00

Specifications

Antigen CD31/PECAM-1
Clone C31.3
Concentration 0.2mg/mL
Immunogen 1990).
Gene Accession No. P16284
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP21520201 Novus Biologicals Canada
NBP21520201MG
0.1mg Each for $329.00
NBP21520202 Novus Biologicals Canada
NBP21520202MG
0.2mg Each for $472.00
Specifications Description & Specifications

Specifications

CD31/PECAM-1
0.2mg/mL
P16284
PBS with 0.05% BSA. Protein G purified.
Primary
Murine
Unlabeled
Purified
C31.3
1990).
5175
Monoclonal
Store at 4°C. Do not freeze.
IgG1, κ
Purified
ELISA,Flow Cytometry,Immunocytochemistry,Immunofluorescence,Immunohistochemistry (Frozen),Immunohistochemistry (Paraffin),Immunoprecipitation,Western Blot