Please login to your online account to display your discounted pricing

Mouse anti-CD31/PECAM-1, Clone: C31.3, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP21520202MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP21520202

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Monoclonal or Polyclonal Monoclonal
Regulatory Status RUO
Isotype IgG1, κ
Antigen CD31/PECAM-1
Cross Reactivity Human
Formulation PBS with 0.05% BSA. Protein G purified.
Conjugate Unlabeled
Primary or Secondary Primary
Format Purified
Storage Requirements Store at 4°C. Do not freeze.
Immunogen 1990).
Quantity 0.2mg
Gene ID (Entrez) 5175
Test Specificity CD31 (PECAM-1) is a transmembrane glycoprotein member of the immunoglobulin supergene family of adhesion molecules. CD31 is expressed by stem cells of the hematopoietic system and is primarily used to identify and concentrate these cells for experimental studies as well as for bone marrow transplantation. Anti-CD31 has shown to be highly specific and sensitive for vascular endothelial cells. Staining of nonvascular tumors (excluding hematopoietic neoplasms) is rare. CD31 MAb reacts with normal, benign, and malignant endothelial cells which make up blood vessel lining. The level of CD31 expression can help to determine the degree of tumor angiogenesis, and a high level of CD31 expression may imply a rapidly growing tumor and potentially a predictor of tumor recurrence.
Purification Method Purified
Clone C31.3
Gene Accession No. P16284
Concentration 0.2mg/mL
Host Species Mouse