Please login to your online account to display your discounted pricing

Mouse anti-CD31/PECAM-1, Clone: C31.3 + JC/70A, Novus Biologicals™

$139.00 - $321.00

Specifications

Antigen CD31/PECAM-1
Clone C31.3 + JC/70A
Concentration 0.2mg/mL
Immunogen Human recombinant CD31 protein (C31.3) and Membrane preparation of a spleen from a patient with hairy cell leukemia (JC/70A)
Gene Accession No. P16284
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23428301

 
novus biologicals canada
NBP23428301ML
0.1mL Each for $139.00

NBP23428305

 
novus biologicals canada
NBP23428305ML
0.5mL Each for $321.00
Description & Specifications

Specifications

Antigen CD31/PECAM-1
Clone C31.3 + JC/70A
Concentration 0.2mg/mL
Immunogen Human recombinant CD31 protein (C31.3) and Membrane preparation of a spleen from a patient with hairy cell leukemia (JC/70A)
Gene Accession No. P16284
Gene ID (Entrez) 5175
Formulation PBS with 0.05% BSA.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Human
Cross Reactivity Primate
Cross Reactivity Rabbit
Isotype IgG1, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting