Please login to your online account to display your discounted pricing

Mouse anti-CD31/PECAM-1, Clone: C31.3 + JC/70A, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23428301ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23428301

  • / Each
 

Description & Specifications

Specifications

Antigen CD31/PECAM-1
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Clone C31.3 + JC/70A
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Primate
Cross Reactivity Rabbit
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. P16284
Host Species Mouse
Immunogen Human recombinant CD31 protein (C31.3) and Membrane preparation of a spleen from a patient with hairy cell leukemia (JC/70A)
Isotype IgG1, κ
Purification Method Purified
Quantity 0.1mL
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 5175
Test Specificity CD31 (PECAM-1) is a transmembrane glycoprotein member of the immunoglobulin supergene family of adhesion molecules. CD31 is expressed by stem cells of the hematopoietic system and is primarily used to identify and concentrate these cells for experimental studies as well as for bone marrow transplantation. Anti-CD31 has shown to be highly specific and sensitive for vascular endothelial cells. Staining of nonvascular tumors (excluding hematopoietic neoplasms) is rare. CD31 MAb reacts with normal, benign, and malignant endothelial cells which make up blood vessel lining. The level of CD31 expression can help to determine the degree of tumor angiogenesis, and a high level of CD31 expression may imply a rapidly growing tumor and potentially a predictor of tumor recurrence. Cross Reacts with Cynomolgus Monkey
Reconstitution Protein A purified