missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mouse anti-CD31/PECAM-1, Clone: C31.3 + JC/70A, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen CD31/PECAM-1
Clone C31.3 + JC/70A
Concentration 1mg/mL
Immunogen Human recombinant CD31 protein (C31.3) and Membrane preparation of a spleen from a patient with hairy cell leukemia (JC/70A)
Gene Accession No. P16284
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP23457801 Novus Biologicals Canada
NBP23457801MG
0.1mg Each for $384.00
NBP23457802 Novus Biologicals Canada
NBP23457802MG
0.2mg Each for $527.00
Specifications Description & Specifications

Specifications

CD31/PECAM-1
1mg/mL
P16284
10mM PBS. Does not contain BSA. Azide Free.
Primary
Murine
Unlabeled
Purified
C31.3 + JC/70A
Human recombinant CD31 protein (C31.3) and Membrane preparation of a spleen from a patient with hairy cell leukemia (JC/70A)
5175
Monoclonal
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG1, κ
Purified
Flow Cytometry,Immunocytochemistry,Immunofluorescence,Immunohistochemistry (Frozen),Immunohistochemistry (Paraffin),Immunoprecipitation,Western Blot