Please login to your online account to display your discounted pricing

Mouse anti-CD300e/LMIR6, Clone: UP-H2, Novus Biologicals™

$274.00 - $329.00

Specifications

Antigen CD300e/LMIR6
Clone UP-H2
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was IREM2.
Gene ID (Entrez) 342510
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14332501

 
novus biologicals canada
NBP14332501MG
0.1mg Each for $329.00

NBP14332503

 
novus biologicals canada
NBP1433250025MG
0.025mg Each for $274.00
Description & Specifications

Specifications

Antigen CD300e/LMIR6
Clone UP-H2
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was IREM2.
Gene ID (Entrez) 342510
Formulation PBS [pH 7.2]
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry
Applications Immunoprecipitation
Applications Western Blotting