Please login to your online account to display your discounted pricing

Mouse anti-CD300e/LMIR6, Clone: UP-H2, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP1433250025MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14332503

  • / Each
 

Description & Specifications

Specifications

Monoclonal or Polyclonal Monoclonal
Applications Flow Cytometry
Applications Immunoprecipitation
Applications Western Blotting
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Human
Antigen CD300e/LMIR6
Clone UP-H2
Concentration 0.5mg/mL
Format Affinity Purified
Quantity 0.025mg
Regulatory Status RUO
Immunogen The immunogen for this antibody was IREM2.
Isotype IgG1
Primary or Secondary Primary
Formulation PBS [pH 7.2]
Purification Method Affinity Purified
Gene ID (Entrez) 342510
Conjugate Unlabeled