Please login to your online account to display your discounted pricing

Mouse anti-CD30/TNFRSF8, Clone: Ber-H2, PE, Novus Biologicals™

$131.00 - $296.00

Specifications

Antigen CD30/TNFRSF8
Clone Ber-H2
Immunogen The immunogen for this antibody is CD30.
Gene ID (Entrez) 943
Monoclonal or Polyclonal Monoclonal
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP200219

 
novus biologicals canada
NBP200219100TESTS
100 Tests Each for $296.00

NBP20021925

 
novus biologicals canada
NBP20021925TESTS
25 Tests Each for $131.00
Description & Specifications

Specifications

Antigen CD30/TNFRSF8
Clone Ber-H2
Immunogen The immunogen for this antibody is CD30.
Gene ID (Entrez) 943
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1
Conjugate PE
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry