Please login to your online account to display your discounted pricing

Mouse anti-CD3, Clone: HIT3a, PE, Novus Biologicals™

$131.00 - $296.00

Specifications

Antigen CD3
Clone HIT3a
Immunogen The immunogen for this antibody was CD3.
Gene ID (Entrez) 916
Formulation Phosphate buffer pH 7.2
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14390325

 
novus biologicals canada
NBP14390325TESTS
25 Tests Each for $131.00

NBP143903

 
novus biologicals canada
NBP143903100TESTS
100 Tests Each for $296.00
Description & Specifications

Specifications

Antigen CD3
Clone HIT3a
Immunogen The immunogen for this antibody was CD3.
Gene ID (Entrez) 916
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Murine
Isotype IgG2a, κ
Conjugate PE
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry