Please login to your online account to display your discounted pricing

Mouse anti-CD3, Clone: HIT3a, Novus Biologicals™

$219.00 - $285.00

Specifications

Antigen CD3
Clone HIT3a
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CD3.
Gene ID (Entrez) 916
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14345501

 
novus biologicals canada
NBP14345501MG
0.1mg Each for $285.00

NBP14345503

 
novus biologicals canada
NBP1434550025MG
0.025mg Each for $219.00
Description & Specifications

Specifications

Antigen CD3
Clone HIT3a
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CD3.
Gene ID (Entrez) 916
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Human
Isotype IgG2a, κ
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry