Please login to your online account to display your discounted pricing

Mouse anti-CD3, Clone: HIT3a, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP1434550025MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14345503

  • / Each
 

Description & Specifications

Specifications

Antigen CD3
Applications Flow Cytometry
Clone HIT3a
Concentration 0.5mg/mL
Conjugate Unlabeled
Format Affinity Purified
Formulation Phosphate buffer pH 7.2
Host Species Murine
Immunogen The immunogen for this antibody was CD3.
Isotype IgG2a, κ
Purification Method Affinity Purified
Quantity 0.025mg
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 916
Target Species Human