Please login to your online account to display your discounted pricing

Mouse anti-CD3, Clone: HIT3a, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14345501MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14345501

  • / Each
 

Description & Specifications

Specifications

Format Affinity Purified
Host Species Mouse
Quantity 0.1mg
Regulatory Status RUO
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 916
Concentration 0.5mg/mL
Antigen CD3
Formulation Phosphate buffer pH 7.2
Applications Flow Cytometry
Immunogen The immunogen for this antibody was CD3.
Isotype IgG2a, κ
Purification Method Affinity Purified
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Clone HIT3a
Conjugate Unlabeled
Cross Reactivity Human