missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mouse anti-CD3, Clone: G4.18, Novus Biologicals™

$219.00 - $340.00

Specifications

Antigen CD3
Clone G4.18
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CD3.
Gene ID (Entrez) 916
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP14345801 Novus Biologicals Canada
NBP14345801MG
0.1mg Each for $285.00
NBP14345805 Novus Biologicals Canada
NBP143458005MG
0.05mg Each for $219.00
NBP1434585 Novus Biologicals Canada
NBP14345805MG
0.5mg Each for $340.00
Specifications Description & Specifications

Specifications

CD3
0.5mg/mL
916
Monoclonal
Store at 4°C. Do not freeze.
IgG3, κ
Affinity Purified
Flow Cytometry,Immunohistochemistry,Immunohistochemistry (Paraffin),Immunoprecipitation,Western Blot
G4.18
The immunogen for this antibody was CD3.
Phosphate buffer pH 7.2
Primary
Murine
Unlabeled
Affinity Purified