missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mouse anti-CD3, Clone: G4.18, Biotin, Novus Biologicals™

$131.00 - $395.00

Specifications

Antigen CD3
Clone G4.18
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CD3.
Gene ID (Entrez) 916
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP14369301 Novus Biologicals Canada
NBP14369301MG
0.1mg Each for $197.00
NBP14369305 Novus Biologicals Canada
NBP143693005MG
0.05mg Each for $131.00
NBP1436935 Novus Biologicals Canada
NBP14369305MG
0.5mg Each for $395.00
Specifications Description & Specifications

Specifications

CD3
0.5mg/mL
916
Monoclonal
Store at 4°C. Do not freeze.
IgG3, κ
Affinity Purified
Flow Cytometry
G4.18
The immunogen for this antibody was CD3.
PBS [pH 7.2]
Primary
Murine
Biotin
Affinity Purified