Please login to your online account to display your discounted pricing

Mouse anti-CD28, Clone: CB28, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23459201MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23459201

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Functional Assay
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Isotype IgG1, κ
Antigen CD28
Cross Reactivity Human
Conjugate Unlabeled
Primary or Secondary Primary
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Monoclonal or Polyclonal Monoclonal
Quantity 0.1mg
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Format Purified
Regulatory Status RUO
Gene ID (Entrez) 940
Reconstitution Protein A purified
Immunogen Recombinant human CD28 protein
Gene Accession No. P10747
Purification Method Purified
Clone CB28
Concentration 1mg/mL
Host Species Mouse