Please login to your online account to display your discounted pricing

Mouse anti-CD28, Clone: C28/75, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen CD28
Clone C28/75
Concentration 1mg/mL
Immunogen Recombinant human CD28 protein
Gene Accession No. P10747
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23457501

 
novus biologicals canada
NBP23457501MG
0.1mg Each for $384.00

NBP23457502

 
novus biologicals canada
NBP23457502MG
0.2mg Each for $527.00
Description & Specifications

Specifications

Antigen CD28
Clone C28/75
Concentration 1mg/mL
Immunogen Recombinant human CD28 protein
Gene Accession No. P10747
Gene ID (Entrez) 940
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Murine
Isotype IgG1, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Functional Assay
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation