Please login to your online account to display your discounted pricing

Mouse anti-CD28, Clone: C28/75, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23457501MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23457501

  • / Each
 

Description & Specifications

Specifications

Antigen CD28
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Host Species Mouse
Applications ELISA
Applications Flow Cytometry
Applications Functional Assay
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Immunogen Recombinant human CD28 protein
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Reconstitution Protein A purified
Clone C28/75
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Gene Accession No. P10747
Isotype IgG1, κ
Purification Method Purified
Quantity 0.1mg
Regulatory Status RUO
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 940
Concentration 1mg/mL